Assay Details
Gene Symbol: SCIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271842.1 | NP_001258771.1 | 2 - 3 | 229 | 119 | |
NM_001319190.1 | NP_001306119.1 | 2 - 3 | 229 | 119 | ||
GenBank mRNA | AK310145.1 | - | 2 - 3 | 229 | 119 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173645.hg.1 | TC1700009556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173650.hg.1 | TC1700009556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009556.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173643.hg.1 | TC1700009556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173647.hg.1 | TC1700009556.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000399600.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000571800.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000574081.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | ENST00000574297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | NM_001271842.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | NM_207103.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173649.hg.1 | TC1700009556.hg.1 | Clariom D TC |
