Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012035.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012035.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012035.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182819.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182819.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182819.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182819.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182819.hg.1 |
TC0600012035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600182820.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182820.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182820.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600182820.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600182820.hg.1 |
TC0600012035.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012035.hg.1 |
ENST00000418518.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012035.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012035.hg.1 |
NR_131248.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012035.hg.1 |
NR_131248.1 |
Refseq |