Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030766.hg.1 |
TC01001987.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030766.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030766.hg.1 |
ENST00000366537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030766.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030765.hg.1 |
TC01001987.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030765.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030765.hg.1 |
ENST00000366537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030765.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189928.hg.1 |
TC0100012219.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189930.hg.1 |
TC0100012219.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000308105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000366537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000470211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
ENST00000486803.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001012970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012219.hg.1 |
NM_001276349.1 |
Refseq |