Assay Details
Gene Symbol: MT1DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003658.2 | 1 - 2 | 209 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000470.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 128258 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 20428 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 760348 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 288284 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3780505 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3231157 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4229 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25223 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46464 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P20477068 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3780505 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3780505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575407 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035389 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035402 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035390 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035412 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035398 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575398 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575397 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035411 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035395 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035389 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035412 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035403 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035395 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575398 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035411 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035402 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575397 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575404 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575407 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035398 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035390 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P200435 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035378 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035389 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899454 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035448 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035374 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035366 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035375 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035406 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035410 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035404 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035408 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035439 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035387 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035442 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035368 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035457 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035451 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035432 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035411 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035413 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035402 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035376 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035382 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035455 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035364 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035436 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035445 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035462 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035453 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035363 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035365 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035403 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035395 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899456 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035409 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035371 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035400 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035441 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035460 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035390 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P200435 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_003658_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7995814 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7995816 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7995815 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7995817 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7995813 | - | - |
Affymetrix Human Genome U219 Array | 11763942_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11761961_s_at | - | - |
Affymetrix Human Genome U219 Array | 11763943_at | - | - |
Affymetrix Human Genome U219 Array | 11763943_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11761961_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11763942_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR16006499.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006499.hg.1 | TC16000470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16006500.hg.1 | ENST00000463480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16006500.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006500.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006500.hg.1 | TC16000470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006502.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006502.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006502.hg.1 | TC16000470.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000334346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000562399.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000567054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000567054.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000568608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000568608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | ENST00000568608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007964.hg.1 | NM_005947.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006501.hg.1 | ENST00000463480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16006501.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006501.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006501.hg.1 | TC16000470.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600158098.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158098.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158098.hg.1 | TC1600007963.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158099.hg.1 | TC1600007963.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600158103.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158103.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158103.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158103.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158103.hg.1 | TC1600007963.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007963.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | ENST00000463480.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | ENST00000486551.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | NR_003658.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | NR_027781.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600158101.hg.1 | TC1600007963.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054623 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054621 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054621 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054623 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 103054 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757867 | AF348999.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1757867 | AF348999.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1757867 | AF348999.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757867 | AF348999.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757867 | AF348999.2 | human_transcriptome |
