Assay Details
| Gene Symbol: LINC01427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110615.1 | 2 - 3 | 199 | 75 | ||
| GenBank mRNA | AI142372.1 | - | 2 - 3 | 199 | 75 | |
| AI147364.1 | - | 1 - 2 | 75 | |||
| BU568468.1 | - | 2 - 3 | 369 | 75 | ||
| CR627206.1 | - | 1 - 2 | 133 | 75 | ||
More Information
Set Membership: |
|

LINC01427
Genomic Map