Assay Details
| Gene Symbol: LOC101929715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110597.1 | 1 - 2 | 67 | |||
| GenBank mRNA | AW303360.1 | - | 3 - 4 | 388 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929715| Gene Symbol: LOC101929715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110597.1 | 1 - 2 | 67 | |||
| GenBank mRNA | AW303360.1 | - | 3 - 4 | 388 | 67 | |
Set Membership: |
|