Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002184.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002184.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002184.hg.1 |
TC10000183.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000185280.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185280.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185280.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185280.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185280.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185280.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185280.hg.1 |
TC1000012435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
ENST00000412114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012435.hg.1 |
NR_026793.1 |
Refseq |