Assay Details
| Gene Symbol: TMCO5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046005.1 | 5 - 6 | 399 | 79 | ||
| GenBank mRNA | DB054691.1 | - | 5 - 6 | 399 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001185.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3350858 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15010499.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010499.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010498.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010498.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010497.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010497.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010495.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010495.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010494.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010494.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010496.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010496.hg.1 | TC15001185.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164656.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164662.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164662.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164662.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164662.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164662.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164662.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164662.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164655.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164643.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164643.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164643.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164652.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164648.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164661.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164661.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164661.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164661.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164661.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164661.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164661.hg.1 | TC1500008962.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500164646.hg.1 | TC1500008962.hg.1 | Clariom D TC |

TMCO5B
Genomic Map