Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172037.4 | NP_742034.1 | 1 - 2 | 984 | 125 | |
| GenBank mRNA | AF456765.1 | - | 1 - 2 | 556 | 125 | |
| AY358270.1 | - | 1 - 2 | 376 | 125 | ||
| BC067131.1 | - | 1 - 2 | 484 | 6153778 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000482.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
metanephros development
in utero embryonic development
retinoic acid biosynthetic process
visual perception
gonad development
neural crest cell development
embryonic camera-type eye development
embryonic forelimb morphogenesis
retinol metabolic process
retinal metabolic process
ear development
nose development
embryonic viscerocranium morphogenesis
oxidation-reduction process
primary lung bud formation
bud elongation involved in lung branching
retinol dehydrogenase activity
NADP-retinol dehydrogenase activity
retinoid metabolic process
metanephros development
in utero embryonic development
retinoic acid biosynthetic process
visual perception
gonad development
neural crest cell development
embryonic camera-type eye development
embryonic forelimb morphogenesis
retinol metabolic process
retinal metabolic process
ear development
nose development
embryonic viscerocranium morphogenesis
oxidation-reduction process
primary lung bud formation
bud elongation involved in lung branching
retinol dehydrogenase activity
NADP-retinol dehydrogenase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08006654.hg.1 | TC08000482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006654.hg.1 | ENST00000240285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006654.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006660.hg.1 | TC08000482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006660.hg.1 | ENST00000240285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006660.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006660.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006656.hg.1 | TC08000482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006656.hg.1 | ENST00000240285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006656.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006655.hg.1 | TC08000482.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006655.hg.1 | ENST00000240285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006655.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154850.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154850.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154850.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154850.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154850.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154850.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154850.hg.1 | TC0800007957.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800154848.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154848.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154848.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154848.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154848.hg.1 | TC0800007957.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154853.hg.1 | TC0800007957.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154847.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154847.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154847.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154847.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154847.hg.1 | TC0800007957.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800154849.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154849.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154849.hg.1 | ENST00000240285.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800154849.hg.1 | NM_172037.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800154849.hg.1 | TC0800007957.hg.1 | Clariom D TC |

RDH10
Genomic Map