Assay Details
| Gene Symbol: LOC100506675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120526.1 | 1 - 2 | 215 | 114 | ||
| GenBank mRNA | DB473712.1 | - | 1 - 2 | 215 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506675| Gene Symbol: LOC100506675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120526.1 | 1 - 2 | 215 | 114 | ||
| GenBank mRNA | DB473712.1 | - | 1 - 2 | 215 | 114 | |
Set Membership: |
|