Assay Details
Gene Symbol: PPP1R42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013626.2 | NP_001013648.1 | 3 - 4 | 440 | 127 | |
GenBank mRNA | BC055413.1 | - | 3 - 4 | 442 | 6614745 | 127 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08002584.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08016625.hg.1 | TC08002584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08016625.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08016625.hg.1 | ENST00000421742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08016625.hg.1 | ENST00000522909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08016625.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08016623.hg.1 | TC08002584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08016623.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08016623.hg.1 | ENST00000421742.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08016623.hg.1 | ENST00000522909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08016623.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187236.hg.1 | TC0800012423.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000518588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000519987.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000520633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521410.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521581.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000521998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | ENST00000523545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012423.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000324682.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000519919.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000521040.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | ENST00000522909.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | NM_001013626.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187237.hg.1 | TC0800012423.hg.1 | Clariom D TC |
