| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018241.hg.1 |
TC08001407.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018241.hg.1 |
ENST00000517562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018241.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011074.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800175320.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800175320.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800175320.hg.1 |
ENST00000517562.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800175320.hg.1 |
NM_001190972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800175320.hg.1 |
TC0800011074.hg.1 |
Clariom D TC |