Assay Details
Gene Symbol: LOC100134368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024453.2 | 2 - 3 | 377 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000012.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 36100 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 29156 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 680165 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 886124 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 900134 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 339300 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 100044 | AF211169.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 50411 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 388827 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 211592 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 2344 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 90311 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 251840 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 202582 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 26671 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 400341 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3408057 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4578 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4577 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 5918 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P03100415 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3408057 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3408057 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16836518 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16836513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387810 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387798 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881699 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16836513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387798 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387810 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881699 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30387788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16836518 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09881681 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P03100415 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03100421 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03100415 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 157522 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 65585 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P20506170 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03100420 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03100415 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03100422 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881709 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881727 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881720 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836529 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881693 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881713 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836536 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881672 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881704 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836518 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881722 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836515 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836527 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881683 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881678 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881723 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881726 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881701 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881706 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836547 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836534 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881689 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881685 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836525 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881715 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881728 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881707 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836526 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836510 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881702 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881711 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881697 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881681 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836542 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836545 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881699 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881679 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881712 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881729 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16836532 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09881695 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03100422 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03100415 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1569270_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000435229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16000238.hg.1 | ENST00000412293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16000238.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16000238.hg.1 | TC16000012.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600144335.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600144335.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600144335.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600144335.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600144335.hg.1 | TC1600006459.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000412293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000435229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | ENST00000457760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006459.hg.1 | NR_024453.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 1569270_at | - | - |
Human Genome U133 Plus 2.0 Array | 1569270_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1569270_at1 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054110 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054110 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 122673 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3246871 | NR_024453.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3246871 | BC028568.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3246871 | NR_024453.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3246871 | BC028568.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246871 | NR_024453.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246871 | BC028568.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246871 | NR_024453.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246871 | BC028568.2 | human_transcriptome |
