| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055987.hg.1 |
TC01003644.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055987.hg.1 |
ENST00000367462.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055987.hg.1 |
ENST00000484105.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055987.hg.1 |
ENST00000534846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055987.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000445957.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000445957.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000445957.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000463404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000463404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000633243.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
ENST00000633243.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055986.hg.1 |
TC01003644.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055986.hg.1 |
ENST00000367462.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055986.hg.1 |
ENST00000484105.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055986.hg.1 |
ENST00000534846.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055986.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226960.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100226960.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226960.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100226960.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226960.hg.1 |
TC0100016755.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010980.hg.1 |
ENST00000452178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000445957.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000445957.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000445957.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000463404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000463404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000631494.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000633243.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
ENST00000633243.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016755.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226959.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100226959.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226959.hg.1 |
ENST00000367462.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100226959.hg.1 |
NM_199051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226959.hg.1 |
TC0100016755.hg.1 |
Clariom D TC |