| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000382252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000427912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000427912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000427912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000427912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000427912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000564837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000614588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000614588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000614588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000614588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000614588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000614588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
ENST00000614588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006652.hg.1 |
NM_001278559.1 |
Refseq |