Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002818.hg.1 |
TC03000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002818.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002818.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002817.hg.1 |
TC03000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002817.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002817.hg.1 |
ENST00000458535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002817.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002816.hg.1 |
TC03000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002816.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002816.hg.1 |
ENST00000458535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002816.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002819.hg.1 |
TC03000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002819.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002819.hg.1 |
ENST00000458535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002819.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148438.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148438.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148438.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148438.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148438.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148438.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148438.hg.1 |
TC0300006961.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300148439.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148439.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148439.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148439.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148439.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148439.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148439.hg.1 |
TC0300006961.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300148440.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148440.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148440.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148440.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148440.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148440.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148440.hg.1 |
TC0300006961.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300148442.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148442.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148442.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148442.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148442.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148442.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148442.hg.1 |
TC0300006961.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
ENST00000458535.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006961.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148441.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148441.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148441.hg.1 |
ENST00000307526.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148441.hg.1 |
NM_178868.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148441.hg.1 |
TC0300006961.hg.1 |
Clariom D TC |