Assay Details
| Gene Symbol: IZUMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031735.2 | NP_001026905.2 | 3 - 4 | 443 | 85 | |
| NM_001039846.1 | NP_001034935.1 | 3 - 4 | 443 | 85 | ||
| XM_005259480.3 | XP_005259537.2 | 3 - 4 | 464 | 85 | ||
| GenBank mRNA | AK294083.1 | - | 3 - 4 | 314 | 85 | |
| AK301957.1 | - | 3 - 4 | 397 | 85 | ||
| AW182303.1 | - | 5 - 6 | 85 | |||
| AY753639.1 | - | 3 - 4 | 402 | 85 | ||
| BC010443.1 | - | 3 - 4 | 382 | 85 | ||
| BC014609.2 | - | 3 - 4 | 397 | 4825119 | 85 | |
| BC040964.1 | - | 3 - 4 | 384 | 85 | ||
| BI827974.1 | - | 3 - 4 | 299 | 85 | ||
| BX378854.2 | - | 3 - 4 | 443 | 85 | ||
| DQ196309.1 | - | 3 - 4 | 430 | 85 | ||
| DQ196310.1 | - | 3 - 4 | 430 | 85 | ||
| DQ424900.1 | - | 3 - 4 | 366 | 85 | ||
More Information
Set Membership: |
|

IZUMO4
Genomic Map