Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015530.hg.1 |
ENST00000330915.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015530.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015530.hg.1 |
TC19001129.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176640.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176640.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176640.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176640.hg.1 |
TC1900009528.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900176640.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176640.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176640.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176638.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176638.hg.1 |
TC1900009528.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900176638.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000330915.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000593649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000595639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
ENST00000610351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009528.hg.1 |
NM_198471.2 |
Refseq |