Assay Details
| Gene Symbol: NBPF18P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125884.1 | - | 6 - 7 | 545 | 156 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000502883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000505709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000506972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012950.hg.1 | ENST00000512915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000413192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | ENST00000432386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | NR_103561.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009980.hg.1 | NR_103561.1 | Refseq |

NBPF18P
Genomic Map