Assay Details
| Gene Symbol: MROH3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ983818.1 | - | 15 - 16 | 2006 | 132 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3242793 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P927416 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011154.hg.1 | ENST00000435735.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 146689 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 208850 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1801284 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1801284 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2073409 | DQ983818.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2073409 | DQ983818.1 | human_transcriptome |

MROH3P
Genomic Map