Assay Details
| Gene Symbol: SNHG21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110096.1 | 1 - 2 | 284 | 100 | ||
| NR_110097.1 | 1 - 2 | 284 | 100 | |||
| NR_110098.1 | 1 - 2 | 284 | 100 | |||
| NR_110099.1 | 1 - 2 | 284 | 100 | |||
| GenBank mRNA | AA284521.1 | - | 2 - 3 | 100 | ||
| BX112407.1 | - | 1 - 2 | 261 | 100 | ||
| CF129490.1 | - | 1 - 2 | 265 | 100 | ||
| CK904397.1 | - | 1 - 2 | 264 | 100 | ||
| DA396616.1 | - | 1 - 2 | 276 | 100 | ||
More Information
Set Membership: |
|

SNHG21
Genomic Map