Assay Details
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC062799.1 | - | 2 - 3 | 815 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000216450.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000393774.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000460005.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000484640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000555189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | ENST00000557453.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007733.hg.1 | NM_194287.2 | Refseq |
