Assay Details
| Gene Symbol: PPP4R1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110773.1 | 2 - 3 | 346 | 83 | ||
| NR_110774.1 | 2 - 3 | 210 | 83 | |||
| GenBank mRNA | BX108181.1 | - | 2 - 3 | 210 | 83 | |
More Information
Set Membership: |
|

PPP4R1-AS1
Genomic Map