| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011282.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011282.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011282.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011282.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011282.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176755.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176755.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176755.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176755.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176755.hg.1 |
TC1000011282.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000176757.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176757.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176757.hg.1 |
TC1000011282.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000176756.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176756.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176756.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176756.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176756.hg.1 |
TC1000011282.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000011282.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011282.hg.1 |
ENST00000444137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011282.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011282.hg.1 |
NR_120669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011282.hg.1 |
NR_120669.1 |
Refseq |