Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008774.hg.1 |
ENST00000339063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008774.hg.1 |
ENST00000409874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008774.hg.1 |
ENST00000554571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008774.hg.1 |
NM_153046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008774.hg.1 |
TC14000810.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008775.hg.1 |
ENST00000339063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008775.hg.1 |
ENST00000409874.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008775.hg.1 |
ENST00000554571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008775.hg.1 |
NM_153046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14008775.hg.1 |
TC14000810.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
ENST00000409874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
ENST00000496087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
ENST00000554571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
NM_153046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
ENST00000409874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
ENST00000496087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
ENST00000554571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
NM_153046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160228.hg.1 |
TC1400008423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
ENST00000409874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
ENST00000496087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
ENST00000554571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
NM_153046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
ENST00000409874.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
ENST00000496087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
ENST00000554571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
NM_153046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400160227.hg.1 |
TC1400008423.hg.1 |
Clariom D TC |