Assay Details
Gene Symbol: LOC100505841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195535.1 | NP_001182464.1 | 1 - 2 | 88 | 60 | |
XM_011543086.2 | XP_011541388.1 | 4 - 5 | 407 | 60 | ||
GenBank mRNA | AI188208.1 | - | 2 - 3 | 60 | ||
BU567915.1 | - | 1 - 2 | 91 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05003442.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001195535.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P662646 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05008018.hg.1 | TC05003442.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05008018.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05008018.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05008021.hg.1 | TC05003442.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05008021.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05008022.hg.1 | TC05003442.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05008022.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05008023.hg.1 | TC05003442.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05008023.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194005.hg.1 | TC0500013222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | ENST00000515596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013222.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194015.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194015.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194015.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194015.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194015.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194015.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194015.hg.1 | TC0500013222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500194016.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194016.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194016.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194016.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194016.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194016.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194016.hg.1 | TC0500013222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | ENST00000509403.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | ENST00000514637.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | ENST00000514887.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | NM_001195535.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194010.hg.1 | TC0500013222.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 231434_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 231434_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231434_at11 | - | - |
Human Genome U95 Set | 88463_at | - | - |
Human Genome U95 Set | 88463_PM_at | - | - |
Human Genome U133 Set | 231434_at | - | - |
Human Genome U133 Set | 231434_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028699 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028699 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 204957 | - | - |
