Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009968.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200169953.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200169953.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200169953.hg.1 |
ENST00000448117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200169953.hg.1 |
NR_126376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200169953.hg.1 |
TC0200009968.hg.1 |
Clariom D TC |