Assay Details
Gene Symbol: RAB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308154.1 | NP_001295083.1 | 1 - 2 | 208 | 66 | |
NM_198686.2 | NP_941959.1 | 1 - 2 | 208 | 66 | ||
GenBank mRNA | AB672631.2 | - | 1 - 2 | 177 | 66 | |
AB672632.1 | - | 1 - 2 | 178 | 66 | ||
AB678452.1 | - | 1 - 2 | 208 | 66 | ||
AB678453.1 | - | 1 - 2 | 191 | 66 | ||
BC040679.2 | - | 1 - 2 | 131 | 4817835 | 66 | |
CN352234.1 | - | 1 - 2 | 209 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14001224.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14013957.hg.1 | ENST00000267512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013957.hg.1 | ENST00000533601.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013957.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14013957.hg.1 | TC14001224.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14013954.hg.1 | ENST00000267512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013954.hg.1 | ENST00000426039.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013954.hg.1 | ENST00000436278.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013954.hg.1 | ENST00000533601.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013954.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14013954.hg.1 | TC14001224.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14013958.hg.1 | ENST00000267512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013958.hg.1 | ENST00000533601.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14013958.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14013958.hg.1 | TC14001224.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000426039.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000554593.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | ENST00000585059.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169646.hg.1 | TC1400009422.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169649.hg.1 | TC1400009422.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | ENST00000267512.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | ENST00000533601.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | NM_001308154.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | NM_198686.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400169651.hg.1 | TC1400009422.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 154797 | - | - |
