Assay Details
| Gene Symbol: RAB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198686.2 | NP_941959.1 | 4 - 5 | 530 | 93 | |
| GenBank mRNA | AB678452.1 | - | 4 - 5 | 530 | 93 | |
| BC040679.2 | - | 4 - 5 | 453 | 4817835 | 93 | |
| BX248046.1 | - | 3 - 4 | 326 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14013946.hg.1 | ENST00000267512.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013946.hg.1 | ENST00000426039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013946.hg.1 | ENST00000436278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013946.hg.1 | ENST00000533601.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013946.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013946.hg.1 | TC14001224.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000267512.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000426039.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000533601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000554593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000555256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | ENST00000585059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_001308154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009422.hg.1 | NM_198686.2 | Refseq |

RAB15
Genomic Map