Assay Details
| Gene Symbol: FAM20C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020223.3 | NP_064608.2 | 2 - 3 | 1012 | 116 | |
| GenBank mRNA | AB545605.1 | - | 2 - 3 | 1012 | 116 | |
| AF533706.1 | - | 2 - 3 | 1012 | 116 | ||
| BC040074.1 | - | 2 - 3 | 868 | 4942737 | 116 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000004.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
positive regulation of bone mineralization
biomineral tissue development
osteoclast maturation
regulation of fibroblast growth factor receptor signaling pathway
positive regulation of osteoblast differentiation
regulation of phosphorus metabolic process
enamel mineralization
odontoblast differentiation
dentinogenesis
protein serine/threonine kinase activity
calcium ion binding
protein binding
ATP binding
manganese ion binding
protein phosphorylation
positive regulation of bone mineralization
biomineral tissue development
osteoclast maturation
regulation of fibroblast growth factor receptor signaling pathway
positive regulation of osteoblast differentiation
regulation of phosphorus metabolic process
enamel mineralization
odontoblast differentiation
dentinogenesis
protein serine/threonine kinase activity
calcium ion binding
protein binding
ATP binding
manganese ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07000023.hg.1 | TC07000004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000023.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000023.hg.1 | ENST00000471328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000023.hg.1 | NM_020223.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000471328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000471328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000020.hg.1 | TC07000004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000020.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000020.hg.1 | NM_020223.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000019.hg.1 | TC07000004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000019.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000019.hg.1 | NM_020223.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000022.hg.1 | TC07000004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000022.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000022.hg.1 | NM_020223.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000025.hg.1 | TC07000004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000025.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000025.hg.1 | NM_020223.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144120.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144120.hg.1 | ENST00000471328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144120.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144120.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144120.hg.1 | ENST00000471328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144120.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144120.hg.1 | TC0700006445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144122.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144122.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144122.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144122.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144122.hg.1 | TC0700006445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144116.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144116.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144116.hg.1 | TC0700006445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144119.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144119.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144119.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144119.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144119.hg.1 | TC0700006445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144117.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144117.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144117.hg.1 | TC0700006445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000313766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000471328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000471328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000477004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000512382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006445.hg.1 | ENST00000515795.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 199772 | - | - |

FAM20C
Genomic Map