Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006064.hg.1 |
ENST00000343998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006064.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006064.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006064.hg.1 |
TC18000575.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006066.hg.1 |
ENST00000343998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006066.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006066.hg.1 |
TC18000575.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006065.hg.1 |
ENST00000343998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006065.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006065.hg.1 |
TC18000575.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800162041.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162041.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162041.hg.1 |
TC1800009034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162041.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162041.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009034.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162042.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162042.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162042.hg.1 |
TC1800009034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162042.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162042.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162040.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162040.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162040.hg.1 |
NM_001044369.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800162040.hg.1 |
TC1800009034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800162040.hg.1 |
ENST00000343998.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162040.hg.1 |
ENST00000400291.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800162040.hg.1 |
NM_001044369.2 |
Refseq |