Assay Details
| Gene Symbol: FAAHP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045483.1 | 3 - 4 | 284 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000586.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3414113 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35321 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3414113 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3414113 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010018.hg.1 | TC01000586.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010018.hg.1 | ENST00000429784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010018.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010017.hg.1 | TC01000586.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010017.hg.1 | ENST00000429784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010017.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010016.hg.1 | TC01000586.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010016.hg.1 | ENST00000429784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010016.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010019.hg.1 | TC01000586.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010019.hg.1 | ENST00000429784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010019.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158411.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158411.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158411.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158411.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158411.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158411.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158411.hg.1 | TC0100008160.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158416.hg.1 | TC0100008160.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158418.hg.1 | TC0100008160.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100158413.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158413.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158413.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158413.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158413.hg.1 | TC0100008160.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000429784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000446499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000606395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | ENST00000607656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008160.hg.1 | NR_045483.1 | Refseq |
| Human Genome U95 Set | 47691_PM_r_at | - | - |
| Human Genome U95 Set | 47691_r_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 169431 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 361497 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3304917 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_3304917 | - | - |

FAAHP1
Genomic Map