Assay Details
| Gene Symbol: LOC151760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038577.1 | - | 1 - 2 | 339 | 5114505 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632325 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632292 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632311 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494726 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632341 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494697 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632310 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494672 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632324 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632345 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632306 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15473981 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632303 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632308 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632336 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632349 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632307 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632290 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02494695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22632347 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62658 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3380252 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3400319 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P342664 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1564837_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1564838_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011973.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011973.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011973.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189112.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189112.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189112.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189112.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189112.hg.1 | TC0300011973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300189113.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189113.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189113.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189113.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189113.hg.1 | TC0300011973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300189114.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189114.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189114.hg.1 | TC0300011973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011973.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011973.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011973.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011973.hg.1 | ENST00000485473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189111.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189111.hg.1 | ENST00000383686.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189111.hg.1 | TC0300011973.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1564837_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564838_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564837_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564838_PM_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564838_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564837_at10 | - | - |
| Human Genome U95 Set | 72175_at | - | - |
| Human Genome U95 Set | 72175_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 305991 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3283369 | - | - |

LOC151760
Genomic Map