Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000438622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000438622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000607074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000607074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
ENST00000607074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
NR_104177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
NR_104177.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010329.hg.1 |
NR_104177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
ENST00000438622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
NR_104177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
ENST00000438622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
NR_104177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170459.hg.1 |
TC0600010329.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000422894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000438622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000438622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000607074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000607074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
ENST00000607074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
NR_104177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
NR_104177.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010329.hg.1 |
NR_104177.1 |
Refseq |