Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007337.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007337.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007337.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007337.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007337.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007337.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149690.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149690.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149690.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149690.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149690.hg.1 |
TC1300007337.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300149688.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149688.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149688.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149688.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149688.hg.1 |
TC1300007337.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300149689.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149689.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149689.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149689.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149689.hg.1 |
TC1300007337.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007337.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007337.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007337.hg.1 |
ENST00000428656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007337.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007337.hg.1 |
NR_046540.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007337.hg.1 |
NR_046540.1 |
Refseq |