Assay Details
| Gene Symbol: RPL29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000992.2 | NP_000983.1 | 1 - 2 | 88 | 154 | |
| GenBank mRNA | BC008926.2 | - | 1 - 2 | 88 | 3835176 | 154 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001444.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC008926.2,NM_000992.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
embryo implantation
viral transcription
cell-cell adhesion
RNA binding
structural constituent of ribosome
heparin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
embryo implantation
viral transcription
cell-cell adhesion
RNA binding
structural constituent of ribosome
heparin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015050.hg.1 | ENST00000432828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010147.hg.1 | ENST00000473590.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007563.hg.1 | ENST00000425197.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011901.hg.1 | ENST00000331979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010647.hg.1 | ENST00000481681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008629.hg.1 | ENST00000421928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008629.hg.1 | ENST00000421928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007733.hg.1 | ENST00000559892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010721.hg.1 | ENST00000494408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010721.hg.1 | ENST00000494408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011221.hg.1 | ENST00000451825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011587.hg.1 | ENST00000478358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012429.hg.1 | ENST00000495837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013451.hg.1 | ENST00000451924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010234.hg.1 | ENST00000463466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010989.hg.1 | ENST00000446953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010989.hg.1 | ENST00000446953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006961.hg.1 | ENST00000496042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006961.hg.1 | ENST00000496042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008499.hg.1 | ENST00000465776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007956.hg.1 | ENST00000431445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009264.hg.1 | ENST00000443998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007734.hg.1 | ENST00000400354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007734.hg.1 | ENST00000400354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007884.hg.1 | ENST00000482584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000484381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008766.hg.1 | ENST00000413677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007312.hg.1 | ENST00000442089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010538.hg.1 | ENST00000440227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000294189.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000294189.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000294189.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000294189.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000294189.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000294189.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000466397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000466397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000466397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000466397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000466397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000466397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000466397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000475248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000475248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000475248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000475248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000475248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000479017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000479017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000479017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000479017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000479017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000479017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000480306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000480306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000480306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000480306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000480306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000480306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000481629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000481629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000481629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000481629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000481629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000481629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000481629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000486565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000486565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000486565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000486565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000486565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000492277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000492277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000492277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000492277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000495383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000495383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000495383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000495383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000495383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000495383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | ENST00000495383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | NM_000992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | NM_000992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | NM_000992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | NM_000992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | NM_000992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011151.hg.1 | NM_000992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008969.hg.1 | ENST00000486611.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011117.hg.1 | ENST00000498718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008766.hg.1 | ENST00000413677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008969.hg.1 | ENST00000486611.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013451.hg.1 | ENST00000451924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009264.hg.1 | ENST00000443998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007884.hg.1 | ENST00000482584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008499.hg.1 | ENST00000465776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010147.hg.1 | ENST00000473590.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011901.hg.1 | ENST00000331979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011117.hg.1 | ENST00000498718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010234.hg.1 | ENST00000463466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007733.hg.1 | ENST00000559892.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015050.hg.1 | ENST00000432828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000470368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000484381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009924.hg.1 | ENST00000496414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010721.hg.1 | ENST00000494408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010721.hg.1 | ENST00000494408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010647.hg.1 | ENST00000481681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007312.hg.1 | ENST00000442089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012429.hg.1 | ENST00000495837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010989.hg.1 | ENST00000446953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010989.hg.1 | ENST00000446953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007563.hg.1 | ENST00000425197.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007956.hg.1 | ENST00000431445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007734.hg.1 | ENST00000400354.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007734.hg.1 | ENST00000400354.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010538.hg.1 | ENST00000440227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011221.hg.1 | ENST00000451825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006961.hg.1 | ENST00000496042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006961.hg.1 | ENST00000496042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011587.hg.1 | ENST00000478358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008629.hg.1 | ENST00000421928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008629.hg.1 | ENST00000421928.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 130747 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 178373 | - | - |

RPL29
Genomic Map