Assay Details
| Gene Symbol: ATP1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001677.3 | NP_001668.1 | 1 - 2 | 220 | 110 | |
| GenBank mRNA | AK290181.1 | - | 1 - 2 | 233 | 110 | |
| AY545986.1 | - | 1 - 2 | 99 | 110 | ||
| BC000006.2 | - | 1 - 2 | 185 | 3506311 | 110 | |
| BQ934151.1 | - | 2 - 3 | 424 | 110 | ||
| BT009787.1 | - | 1 - 2 | 99 | 110 | ||
| CR407616.1 | - | 1 - 2 | 99 | 110 | ||
| U16799.1 | - | 1 - 2 | 144 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001473.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of defense response to virus by host
cellular calcium ion homeostasis
cellular sodium ion homeostasis
cell adhesion
potassium ion import
establishment or maintenance of transmembrane electrochemical gradient
regulation of gene expression
regulation of cardiac muscle contraction by calcium ion signaling
cellular potassium ion homeostasis
positive regulation of ATPase activity
ion transmembrane transport
sodium ion export from cell
protein transport into plasma membrane raft
ATP metabolic process
protein stabilization
leukocyte migration
relaxation of cardiac muscle
cardiac muscle contraction
protein localization to plasma membrane
membrane repolarization
membrane repolarization during cardiac muscle cell action potential
cell communication by electrical coupling involved in cardiac conduction
ATP hydrolysis coupled transmembrane transport
mitophagy in response to mitochondrial depolarization
xenophagy
positive regulation of potassium ion transmembrane transporter activity
positive regulation of sodium ion export from cell
positive regulation of calcium:sodium antiporter activity
positive regulation of potassium ion import
regulation of cardiac conduction
potassium ion import across plasma membrane
ATPase activator activity
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
protein C-terminus binding
drug binding
ATPase activity
MHC class II protein complex binding
potassium ion binding
sodium ion binding
ATPase binding
response to hypoxia
positive regulation of defense response to virus by host
cellular calcium ion homeostasis
cellular sodium ion homeostasis
cell adhesion
potassium ion import
establishment or maintenance of transmembrane electrochemical gradient
regulation of gene expression
regulation of cardiac muscle contraction by calcium ion signaling
cellular potassium ion homeostasis
positive regulation of ATPase activity
ion transmembrane transport
sodium ion export from cell
protein transport into plasma membrane raft
ATP metabolic process
protein stabilization
leukocyte migration
relaxation of cardiac muscle
cardiac muscle contraction
protein localization to plasma membrane
membrane repolarization
membrane repolarization during cardiac muscle cell action potential
cell communication by electrical coupling involved in cardiac conduction
ATP hydrolysis coupled transmembrane transport
mitophagy in response to mitochondrial depolarization
xenophagy
positive regulation of potassium ion transmembrane transporter activity
positive regulation of sodium ion export from cell
positive regulation of calcium:sodium antiporter activity
positive regulation of potassium ion import
regulation of cardiac conduction
potassium ion import across plasma membrane
ATPase activator activity
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
protein C-terminus binding
drug binding
ATPase activity
MHC class II protein complex binding
potassium ion binding
sodium ion binding
ATPase binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01022837.hg.1 | TC01001473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022837.hg.1 | ENST00000367815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022837.hg.1 | ENST00000367816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022837.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022843.hg.1 | TC01001473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022843.hg.1 | ENST00000367813.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022843.hg.1 | ENST00000367815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022843.hg.1 | ENST00000367816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022843.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022843.hg.1 | ENST00000499679.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022843.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022838.hg.1 | TC01001473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022838.hg.1 | ENST00000367815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022838.hg.1 | ENST00000367816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022838.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007362.hg.1 | ENST00000509767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177592.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177592.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177592.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177592.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177592.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177592.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177592.hg.1 | TC0100010543.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177593.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177593.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177593.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177593.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177593.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177593.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177593.hg.1 | TC0100010543.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177595.hg.1 | TC0100010543.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007362.hg.1 | ENST00000509767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177591.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177591.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177591.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177591.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177591.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177591.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177591.hg.1 | TC0100010543.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367815.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000367816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | ENST00000494797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010543.hg.1 | NM_001677.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 199586 | - | - |

ATP1B1
Genomic Map