Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005297.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005297.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005297.hg.1 |
TC10000445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005296.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005296.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005296.hg.1 |
TC10000445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005298.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005298.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005298.hg.1 |
TC10000445.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000154145.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154145.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154145.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154145.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154145.hg.1 |
TC1000007976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000154148.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154148.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154148.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154148.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154148.hg.1 |
TC1000007976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000154147.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154147.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154147.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154147.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154147.hg.1 |
TC1000007976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000154146.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154146.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154146.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154146.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154146.hg.1 |
TC1000007976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
ENST00000373115.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007976.hg.1 |
NM_004273.4 |
Refseq |