Assay Details
Gene Symbol: CGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020770.2 | NP_065821.1 | 13 - 14 | 2717 | 64 | |
XM_005245365.4 | XP_005245422.1 | 13 - 14 | 2729 | 64 | ||
GenBank mRNA | AB037740.1 | - | 13 - 14 | 2690 | 64 | |
AF263462.1 | - | 13 - 14 | 2724 | 64 | ||
AK290007.1 | - | 13 - 14 | 2690 | 64 | ||
BC146657.1 | - | 12 - 13 | 2995 | 40147940 | 64 | |
BC150604.1 | - | 12 - 13 | 2596 | 9056974 | 64 | |
BC152445.1 | - | 13 - 14 | 2690 | 8862573 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001211.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01018781.hg.1 | TC01001211.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018781.hg.1 | ENST00000271636.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018781.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000502442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000502442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000505188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | ENST00000505188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100172183.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100172183.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100172183.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100172183.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100172183.hg.1 | TC0100009958.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000271636.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000416743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000427934.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000464886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000467998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000473377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000502442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000502442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000505188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | ENST00000505188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009958.hg.1 | NM_020770.2 | Refseq |
