Assay Details
Gene Symbol: PPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002703.4 | NP_002694.3 | 3 - 4 | 562 | 71 | |
XM_011534387.1 | XP_011532689.1 | 3 - 4 | 339 | 71 | ||
GenBank mRNA | AK222759.1 | - | 3 - 4 | 501 | 71 | |
AK290942.1 | - | 3 - 4 | 417 | 71 | ||
AK312297.1 | - | 3 - 4 | 538 | 71 | ||
BC004200.2 | - | 3 - 4 | 434 | 3533162 | 71 | |
D13757.1 | - | 3 - 4 | 524 | 71 | ||
U00238.1 | - | 3 - 4 | 453 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001220.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
kidney development
purine nucleotide biosynthetic process
'de novo' IMP biosynthetic process
glutamine catabolic process
lactation
purine nucleobase biosynthetic process
nucleoside metabolic process
purine ribonucleoside monophosphate biosynthetic process
organ regeneration
cellular response to insulin stimulus
cellular response to drug
protein homotetramerization
maternal process involved in female pregnancy
amidophosphoribosyltransferase activity
metal ion binding
4 iron, 4 sulfur cluster binding
G1/S transition of mitotic cell cycle
kidney development
purine nucleotide biosynthetic process
'de novo' IMP biosynthetic process
glutamine catabolic process
lactation
purine nucleobase biosynthetic process
nucleoside metabolic process
purine ribonucleoside monophosphate biosynthetic process
organ regeneration
cellular response to insulin stimulus
cellular response to drug
protein homotetramerization
maternal process involved in female pregnancy
amidophosphoribosyltransferase activity
metal ion binding
4 iron, 4 sulfur cluster binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04016694.hg.1 | TC04001220.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04016694.hg.1 | ENST00000264220.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016694.hg.1 | ENST00000507648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04016694.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400173350.hg.1 | TC0400010770.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000264220.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000425339.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000507724.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | ENST00000510643.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010770.hg.1 | NM_002703.4 | Refseq |
