Assay Details
Gene Symbol: SLC22A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003058.3 | NP_003049.2 | 5 - 6 | 1127 | 134 | |
GenBank mRNA | AB075951.1 | - | 5 - 6 | 1131 | 134 | |
AK290787.1 | - | 5 - 6 | 1031 | 134 | ||
BC039899.1 | - | 5 - 6 | 1008 | 5186229 | 134 | |
X98333.1 | - | 5 - 6 | 1097 | 134 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002279.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
drug transmembrane transport
neurotransmitter secretion
body fluid secretion
organic cation transport
quaternary ammonium group transport
choline transport
dopamine transport
neurotransmitter biosynthetic process
histamine transport
neurotransmitter transporter activity
dopamine transmembrane transporter activity
steroid binding
organic cation transmembrane transporter activity
choline transmembrane transporter activity
quaternary ammonium group transmembrane transporter activity
drug transmembrane transport
neurotransmitter secretion
body fluid secretion
organic cation transport
quaternary ammonium group transport
choline transport
dopamine transport
neurotransmitter biosynthetic process
histamine transport
neurotransmitter transporter activity
dopamine transmembrane transporter activity
steroid binding
organic cation transmembrane transporter activity
choline transmembrane transporter activity
quaternary ammonium group transmembrane transporter activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06028593.hg.1 | TC06002279.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06028593.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028593.hg.1 | ENST00000366953.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028593.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028593.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000366952.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000366953.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000486916.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | ENST00000491092.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | NM_003058.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600194993.hg.1 | TC0600013775.hg.1 | Clariom D TC |
