Assay Details
| Gene Symbol: FKBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF100751.1 | - | 3 - 4 | 574 | 88 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000233092.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000412612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000419184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000424785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000434643.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000435079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000464248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | ENST00000470945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_001135212.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015082.hg.1 | NM_181342.2 | Refseq |

FKBP7
Genomic Map