Assay Details
| Gene Symbol: C8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023080.2 | NP_075568.1 | 1 - 2 | 60 | 66 | |
| GenBank mRNA | AK024642.1 | - | 1 - 2 | 58 | 66 | |
| DA467518.1 | - | 1 - 2 | 60 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000867.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK024642.1,NM_023080.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08011529.hg.1 | TC08000867.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011529.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011529.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011531.hg.1 | TC08000867.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011531.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011531.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163818.hg.1 | TC0800009301.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163819.hg.1 | TC0800009301.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800163811.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163811.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163811.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163811.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163811.hg.1 | TC0800009301.hg.1 | Clariom D TC |

C8orf33
Genomic Map