| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000594473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000594473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900197993.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197993.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197993.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900197993.hg.1 |
TC1900011587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900197993.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197993.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197993.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000594473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000594473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000596248.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
ENST00000600397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011587.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900197992.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197992.hg.1 |
NM_024762.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900197992.hg.1 |
TC1900011587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900197992.hg.1 |
ENST00000391701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900197992.hg.1 |
NM_024762.3 |
Refseq |