| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001954.hg.1 |
ENST00000306243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001954.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001954.hg.1 |
TC15000286.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001956.hg.1 |
ENST00000306243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001956.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001956.hg.1 |
TC15000286.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001955.hg.1 |
ENST00000306243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001955.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001955.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001955.hg.1 |
TC15000286.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001957.hg.1 |
ENST00000306243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001957.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001957.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001957.hg.1 |
TC15000286.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001951.hg.1 |
ENST00000306243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001951.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001951.hg.1 |
TC15000286.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001953.hg.1 |
ENST00000306243.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001953.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001953.hg.1 |
TC15000286.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500148465.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148465.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148465.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148465.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148465.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148465.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148465.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500148460.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148460.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148460.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500148466.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148466.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148466.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148466.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148466.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500148468.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148468.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148468.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148468.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148468.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148468.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148468.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006959.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148467.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148467.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148467.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148467.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148467.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148467.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148467.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500148462.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148462.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148462.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148462.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148462.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500148463.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148463.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148463.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148463.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148463.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500148464.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148464.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148464.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148464.hg.1 |
ENST00000306243.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148464.hg.1 |
ENST00000559991.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500148464.hg.1 |
NM_130468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500148464.hg.1 |
TC1500006959.hg.1 |
Clariom D TC |