Assay Details
| Gene Symbol: LOC142937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008131.2 | - | 1 | 620 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401857 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046922 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401862 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046892 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046916 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046898 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046900 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27401833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046880 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046908 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07046893 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3544880 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P402164 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 14546 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24788 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3544880 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P402164 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P402164 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1552986_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1552986_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1552986_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | XM_495796_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | XM_938400_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1552986_at7 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 155964 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1701230 | BC008131.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1701230 | BC008131.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1701230 | BC008131.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1701230 | BC008131.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 3930520 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_32173 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1701230 | BC008131.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1701230 | BC008131.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1701230 | BC008131.2 | human_transcriptome |

LOC142937
Genomic Map