Assay Details
| Gene Symbol: SLC39A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144564.4 | NP_653165.2 | 1 - 2 | 135 | 109 | |
| NM_213568.1 | NP_998733.1 | 1 - 2 | 135 | 109 | ||
| GenBank mRNA | AK002044.1 | - | 1 - 2 | 98 | 109 | |
| BC020571.2 | - | 1 - 2 | 103 | 109 | ||
| BG719479.1 | - | 1 - 2 | 112 | 109 | ||
| BQ898834.1 | - | 211 | 109 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001035.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK002044.1,BC020571.2,NM_144564.4,NM_213568.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19014113.hg.1 | ENST00000455372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014113.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014113.hg.1 | TC19001035.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014115.hg.1 | ENST00000269740.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014115.hg.1 | ENST00000455372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014115.hg.1 | ENST00000545664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014115.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014115.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014115.hg.1 | TC19001035.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014116.hg.1 | ENST00000269740.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014116.hg.1 | ENST00000455372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014116.hg.1 | ENST00000545664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014116.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014116.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014116.hg.1 | TC19001035.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014114.hg.1 | ENST00000269740.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014114.hg.1 | ENST00000455372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014114.hg.1 | ENST00000545664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014114.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014114.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014114.hg.1 | TC19001035.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014112.hg.1 | ENST00000455372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014112.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014112.hg.1 | TC19001035.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014118.hg.1 | ENST00000269740.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014118.hg.1 | ENST00000455372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014118.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014118.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014118.hg.1 | TC19001035.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | TC1900011843.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203570.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | TC1900011843.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203576.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203579.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203579.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203579.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203579.hg.1 | TC1900011843.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203579.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203579.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203579.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203568.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203568.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203568.hg.1 | TC1900011843.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203568.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203568.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000589363.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011843.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | TC1900011843.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000269740.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000545664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000589166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | ENST00000590875.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | NM_144564.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203573.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203569.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203569.hg.1 | NM_213568.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203569.hg.1 | TC1900011843.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203569.hg.1 | ENST00000455372.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203569.hg.1 | NM_213568.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 143461 | - | - |

SLC39A3
Genomic Map