Assay Details
Gene Symbol: HUS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148959.3 | NP_683762.2 | 1 | 177 | 109 | |
GenBank mRNA | AF508547.1 | - | 1 | 157 | 109 | |
BC101697.1 | - | 1 | 177 | 8069203 | 109 | |
BC101699.1 | - | 1 | 177 | 8069205 | 109 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001208.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689310 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689314 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689292 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04502875 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689311 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04502876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24689293 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04502892 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04502885 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P133739 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P423974 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P133739 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 1718 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 20047 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 31741 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P133739 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P133739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689310 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502879 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689310 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502879 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04502883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24689303 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P423974 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P133739 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502891 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502888 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502874 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502876 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502886 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502878 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502881 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502879 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04502883 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15776534 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15776532 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04502877 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15776535 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04502876 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04502878 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15776530 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15776531 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04502880 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15776533 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04502879 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P423974 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P133739 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502878 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502886 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502890 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502893 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502881 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502883 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502888 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15776531 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502895 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502876 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502879 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04502874 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_148959_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123560 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123561 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123559 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8123558 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553660_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553661_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11738299_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738299_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0600010383.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010383.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010383.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010383.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010383.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010383.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06014408.hg.1 | TC06001208.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06014408.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06014408.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600170716.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600170716.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600170716.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600170716.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600170716.hg.1 | TC0600010383.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600010383.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010383.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010383.hg.1 | ENST00000380907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010383.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010383.hg.1 | NM_148959.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010383.hg.1 | NM_148959.3 | Refseq |
Human Genome U133 Plus 2.0 Array | 1553661_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553661_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553660_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553660_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_148959_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553660_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553661_a_at5 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029305 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029305 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 227426 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1706410 | BC101697.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1706410 | BC101699.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1706410 | AF508547.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31077214-S | BC101697.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31077214-S | BC101699.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31077214-S | NM_148959.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31077214-S | AF508547.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31077214-S | AI446694.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_137519-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0003450605 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0003450605 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5900184 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_137519 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1706410 | AI446694.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1706410 | BC101697.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1706410 | BC101699.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1706410 | NM_148959.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1706410 | AF508547.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1706410 | AI446694.1 | human_transcriptome |
