Assay Details
Gene Symbol: GPR156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168271.1 | NP_001161743.1 | 2 - 3 | 427 | 109 | |
NM_153002.2 | NP_694547.2 | 2 - 3 | 427 | 109 | ||
XM_005247165.2 | XP_005247222.1 | 3 - 4 | 704 | 109 | ||
XM_017005795.1 | XP_016861284.1 | 3 - 4 | 740 | 109 | ||
XM_017005796.1 | XP_016861285.1 | 3 - 4 | 749 | 109 | ||
XM_017005797.1 | XP_016861286.1 | 3 - 4 | 704 | 109 | ||
GenBank mRNA | AF488739.1 | - | 2 - 3 | 230 | 109 | |
BC113701.1 | - | 2 - 3 | 250 | 8322753 | 109 | |
BC143606.1 | - | 2 - 3 | 250 | 9052119 | 109 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001689.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03030472.hg.1 | TC03001689.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03030472.hg.1 | ENST00000315843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03030472.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03030472.hg.1 | ENST00000464295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03030472.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03030472.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190340.hg.1 | TC0300012140.hg.1 | Clariom D TC |
