Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030466.hg.1 |
TC03001689.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030466.hg.1 |
ENST00000315843.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030466.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030466.hg.1 |
ENST00000464295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030466.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030466.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190334.hg.1 |
TC0300012140.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000315843.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000461057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000464295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000481907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
ENST00000495912.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_001168271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012140.hg.1 |
NM_153002.2 |
Refseq |